Glyoxylic acid monohydrate for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

Synonyms: 2,2-Dihydroxyacetic acid, Formylformic acid monohydrate, Oxoacetic acid monohydrate, 2-Oxoethanoic acid monohydrate

M-Clarity™ quality level = MQ200

8.04107.0025 8.04107.0100 8.04107.0250 8.04107.0025 8.04107.0100 8.04107.0250
EM8.04107.0025CS 8727.92 CAD
EM8.04107.0025 EM8.04107.0100 EM8.04107.0250 CA8.04107.0025 CA8.04107.0100 CA8.04107.0250
Glyoxylic acid monohydrate for synthesis, Sigma-Aldrich®
Glyoxylic acid monohydrate
Formula: C₂H₂O₃·1H₂O
MW: 92.05 g/mol
MDL Number: MFCD00127974
CAS Number: 563-96-2
EINECS: 206-058-5
UN: 3261
ADR: 8 III

Order Now

Specification Test Results

Assay(acidimetric) ≥96.5%
Identity( IR) passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR