1,3-Dioxolane stabilized for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger


M-Clarity™ quality level = MQ200

8.03553.0100 8.03553.0500 8.03553.0100 8.03553.0500
EM8.03553.0100CS 1433.06 CAD
EM8.03553.0100 EM8.03553.0500 CA8.03553.0100 CA8.03553.0500
1,3-Dioxolane stabilized for synthesis, Sigma-Aldrich®
1,3-Dioxolane
Formula: C₃H₆O₂
MW: 74.08 g/mol
Boiling Pt: 75 °C (1013 hPa)
Melting Pt: –95 °C
Density: 1.066 g/cm³ (15 °C)
MDL Number: MFCD00003207
CAS Number: 646-06-0
EINECS: 211-463-5
UN: 1166
ADR: 3,II

Order Now

Specification Test Results

Solubility (20 °C) soluble
Melting Point -26 °C
Molar Mass 74.08 g/mol
Boiling Point 74 - 75 °C (1013 hPa)
Vapor Pressure 133 hPa (20 °C)
Flash Point 2 °C
Refractive Index 1.4 (20 °C, 589 nm)
Explosion Limit 2.1 - 20.5 %(V)
Density 1.065 g/cm3 (20 °C)
Ignition Point 274 °C
Hazard Class 3
Hazard Class Description Flammable Liquid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR