Dimethyl carbonate for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
|
Danger
|
M-Clarity™ quality level = MQ200
8.03525.1000
8.03525.2500
8.03525.1000
8.03525.2500
EM8.03525.1000CS
1148.66
CAD
EM8.03525.1000
EM8.03525.2500
CA8.03525.1000
CA8.03525.2500
Dimethyl carbonate for synthesis, Sigma-Aldrich®
Dimethyl carbonate
Formula:
C₃H₆O₃ MW: 90.08 g/mol Boiling Pt: 90 Melting Pt: 0.5…4.7 °C Density: 1.07 g/cm³ (20 °C) Flash Pt: 18 Storage Temperature: Ambient |
MDL Number:
MFCD00008420 CAS Number: 616-38-6 EINECS: 210-478-4 UN: 1161 ADR: 3 II Merck Index: 12,06117 |
Specification Test Results
Solubility | 139 g/l (20 °C) |
Melting Point | 0.5 - 4.7 °C |
Molar Mass | 90.08 g/mol |
Boiling Point | 90 °C (1016 hPa) |
Vapor Pressure | 53 hPa (20 °C) |
Flash Point | 16.7 °C |
Explosion Limit | 4.22 - 12.87 %(V) |
Density | 1.07 g/cm3 (20 °C) |
pH | (H2O, 20 °C) no data available |
Ignition Point | 458 °C |
Hazard Class | 3 |
Hazard Class Description | Flammable Liquid |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...