1-Dodecanol ≥98% for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning


M-Clarity™ quality level = MQ200

8.03462.0100 8.03462.1000 8.03462.0100 8.03462.1000
EM8.03462.0100CS 2449 CAD
EM8.03462.0100 EM8.03462.1000 CA8.03462.0100 CA8.03462.1000
1-Dodecanol ≥98% for synthesis, Sigma-Aldrich®
1-Dodecanol
Formula: H₃C(CH₂)₁₀CH₂OH
Boiling Pt: 260 °C (1013 hPa)
Melting Pt: 22…24 °C
Density: 0.81976 g/cm³ (20 °C)
Flash Pt: 119 °C
Storage Temperature: Ambient
MDL Number: MFCD00004753
CAS Number: 112-53-8
EINECS: 203-982-0
UN: 3077
ADR: 9,III
Merck Index: 13,03439

Order Now

Specification Test Results

Melting Point 21-24°C
Hazard Class 9
Hazard Class Description Miscellaneous Hazardous Material


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR