2-Diethylaminoethanol ≥99.0% (by GC) for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

Synonyms: 2-(Diethylamino)-ethanolN,N,-DiethylethanolamineN,N-Diethylethanolamine

M-Clarity™ quality level = MQ200

8.03273.9027 8.03273.9027
CA8.03273.9027EA 1574.15 CAD
CA8.03273.9027 EM8.03273.9027
2-Diethylaminoethanol ≥99.0% (by GC) for synthesis, Sigma-Aldrich®
2-Diethylaminoethanol
Formula: C₆H₁₅NO
MW: 117.19 g/mol
Boiling Pt: 163 °C (1013 hPa)
Melting Pt: –68 °C
Density: 0.88 g/cm3 (20 °C)
Flash Pt: 52 °C
MDL Number: MFCD00002850
CAS Number: 100-37-8
UN: 2686
ADR: 8.3,II
Merck Index: 13,03139

Order Now

Specification Test Results

Solubility (20 °C) soluble
Melting Point -70 °C
Molar Mass 117.19 g/mol
Boiling Point 163 °C (1013 hPa)
Vapor Pressure 1.9 hPa (20 °C)
Flash Point 51 °C
Refractive Index 1.4414 (20 °C, 589 nm)
Explosion Limit 0.7 - 10.1 %(V)
Density 0.88 g/cm3 (25 °C)
pH 11.5 (100 g/l, H2O, 20 °C)
Ignition Point 270 °C
Hazard Class 8
Hazard Class Description Corrosive Material

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR