Di-isopropanolamine for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
|
Warning
|
Synonyms:
1,1'-Iminodipropan-2-ol, Bis(2-hydroxypropyl)amine, 1,1-Iminodi-2-propanol, 1-[(2-Hydroxypropyl)amino]propan-2-ol
M-Clarity™ quality level = MQ200
8.03264.0100
8.03264.1000
8.03264.0100
8.03264.1000
EM8.03264.0100CS
11821.56
CAD
EM8.03264.0100
EM8.03264.1000
CA8.03264.0100
CA8.03264.1000
Di-isopropanolamine for synthesis, Sigma-Aldrich®
Di-isopropanolamine
Formula:
C₆H₁₅NO₂ Storage Temperature: Ambient |
MDL Number:
MFCD00004531 CAS Number: 110-97-4 EINECS: 203-820-9 |
Specification Test Results
Solubility | 870 g/l (20 °C) |
Melting Point | 38 - 44 °C |
Molar Mass | 133.19 g/mol |
Boiling Point | 249 °C (1013 hPa) |
Vapor Pressure | 0.02 hPa (20 °C) |
Flash Point | 135 °C |
Refractive Index | 1.4702 (16 °C, 589 nm) |
Explosion Limit | 1.6 - 8.0 %(V) |
Density | 0.99 g/cm3 (20 °C) |
pH | 11.4 (100 g/l, H2O, 20 °C) |
Ignition Point | 290 °C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...