1,4-Dichlorobenzene for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning

Synonyms: p-Dichlorobenzene

M-Clarity™ quality level = MQ200

8.03226.0100 8.03226.1000 8.03226.2500 8.03226.0100 8.03226.1000 8.03226.2500
EM8.03226.0100CS 9772.3 CAD
EM8.03226.0100 EM8.03226.1000 EM8.03226.2500 CA8.03226.0100 CA8.03226.1000 CA8.03226.2500
1,4-Dichlorobenzene for synthesis, Sigma-Aldrich®
1,4-Dichlorobenzene
Formula: C₆H₄Cl₂
MW: 147 g/mol
Boiling Pt: 173.4 °C (1013 hPa)
Melting Pt: 48…50 °C
Density: 1.458 g/cm³ (20 °C)
Flash Pt: 65 °C
Storage Temperature: Ambient
MDL Number: MFCD00000604
CAS Number: 106-46-7
EINECS: 203-400-5
UN: 3077
ADR: 9,III
Merck Index: 13,03082

Order Now

Specification Test Results

Solubility 0.08 g/l (25 °C)
Melting Point 52 - 54 °C
Molar Mass 147 g/mol
Bulk Density 800 kg/m3
Boiling Point 173 - 175 °C (1013 hPa)
Vapor Pressure 0.8 hPa (20 °C)
Flash Point 66 °C
Refractive Index 1.5267 (70 °C, 589 nm)
Explosion Limit 1.7 - 5.9 %(V)
Density 1.23 g/cm3 (70 °C)
pH 7 (0.06 g/l, H2O, 20 °C)
Ignition Point 500 °C
Saturation concentration 5 g/m3 (20 °C) Air
Hazard Class 9
Hazard Class Description Miscellaneous Hazardous Material

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR