Dibutylamine for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger


M-Clarity™ quality level = MQ200

8.03222.0100 8.03222.2500 8.03222.0100 8.03222.2500
EM8.03222.0100CS 936.94 CAD
EM8.03222.0100 EM8.03222.2500 CA8.03222.0100 CA8.03222.2500
Dibutylamine for synthesis, Sigma-Aldrich®
Dibutylamine
Formula: C₈H₁₉N
Boiling Pt: 159 °C (1013 hPa)
Melting Pt: –62 °C
Density: 0.759 g/cm³ (20 °C)
Flash Pt: 43 °C
Storage Temperature: Ambient
MDL Number: MFCD00009429
CAS Number: 111-92-2
EINECS: 203-921-8
UN: 2248
ADR: 8,II
Merck Index: 13,03058
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b0d7d86589224f8565e12dc8e6b30a8de923393f&vid=d2cc023db25785e54555fc105aa8436e65fd9cbc&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Solubility 4.05 g/l (25 °C)
Melting Point -62 °C
Molar Mass 129.24 g/mol
Boiling Point 160 °C (1013 hPa)
Vapor Pressure 2.3 hPa (20 °C)
Flash Point 39 °C DIN 51755 Part 1
Refractive Index 1.4168 (20 °C, 589 nm)
Explosion Limit 0.6 - 6.8 %(V)
Density 0.76 g/cm3 (20 °C)
pH 11.1 (1 g/l, H2O, 20 °C)
Ignition Point 260 °C DIN 51794
Hazard Class 8
Hazard Class Description Corrosive Material


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR