Benzyl(dimethyl)amine for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

Synonyms: DMBA, N,N-Dimethylbenzylamine, N-Benzyldimethylamine, BDMA

M-Clarity™ quality level = MQ200

8.03206.0250 8.03206.1000 8.03206.0250 8.03206.1000
EM8.03206.0250CS 996.98 CAD
EM8.03206.0250 EM8.03206.1000 CA8.03206.0250 CA8.03206.1000
Benzyl(dimethyl)amine for synthesis, Sigma-Aldrich®
Benzyl(dimethyl)amine
Formula: C₆H₅CH₂N(CH₃)₂
MW: 135.21 g/mol
Boiling Pt: 183
Melting Pt: -75
Density: 0.915 g/cm³ (20 °C)
Flash Pt: 53
Storage Temperature: Ambient
MDL Number: MFCD00008329
CAS Number: 103-83-3
EINECS: 203-149-1
UN: 2619
ADR: 8 (3) II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=90635bbe74e3372ed4bfbd8d750b25afd50f77a5&vid=7044105a7e7fbce7e00f1d15d7a4e017a1b9d418&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Solubility 8 g/l (20 °C)
Melting Point -75 °C
Molar Mass 135.21 g/mol
Boiling Point 180 - 183 °C (1013 hPa)
Vapor Pressure 2.4 hPa (20 °C)
Flash Point 55 °C
Refractive Index 1.5011 (20 °C, 589 nm)
Explosion Limit 0.9 - 6.3 %(V)
Density 0.90 g/cm3 (20 °C)
pH 10 (10 g/l, H2O, 20 °C) (saturated solution)
Ignition Point 410 °C
Hazard Class 8
Hazard Class Description Corrosive Material


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR