Diethylene glycol monobutyl ether for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning

Synonyms: 2-(2-Butoxyethoxy)ethanol, Diethylene glycol mono-n-butyl ether, Butyldigol

M-Clarity™ quality level = MQ200

8.03129.1000 8.03129.1000 8.03129.6010 8.03129.9025
EM8.03129.1000CS 505.6 CAD
EM8.03129.1000 CA8.03129.1000 CA8.03129.6010 CA8.03129.9025
Diethylene glycol monobutyl ether for synthesis, Sigma-Aldrich®
Diethylene glycol monobutyl ether
Formula: CH₃(CH₂)₃OCH₂CH₂OCH₂CH₂OH
MW: 162.23 g/mol
Boiling Pt: 230.4
Melting Pt: -68
Density: 0.954 g/cm³ (20 °C)
Flash Pt: 100
Storage Temperature: Ambient
MDL Number: MFCD00002881
CAS Number: 112-34-5
EINECS: 203-961-6
Merck Index: 13,01556

Order Now

Specification Test Results

Solubility (20 °C) soluble
Melting Point -68 °C
Molar Mass 162.23 g/mol
Boiling Point 226 - 234 °C (1013 hPa)
Vapor Pressure 0.1 hPa (20 °C)
Flash Point 105 °C
Refractive Index 1.4258 (27 °C, 589 nm)
Explosion Limit 0.7 - 5.3 %(V)
Density 0.95 g/cm3 (20 °C)
pH (H2O, 20 °C) neutral
Ignition Point 225 °C
Viscosity kinematic 6.2 mm2/s (20 °C)
Evaporation number 1200


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR