Diethylene glycol monomethyl ether for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning


M-Clarity™ quality level = MQ200

8.03128.1000 8.03128.2500 8.03128.1000 8.03128.2500
EM8.03128.1000CS 646.22 CAD
EM8.03128.1000 EM8.03128.2500 CA8.03128.1000 CA8.03128.2500
Diethylene glycol monomethyl ether for synthesis, Sigma-Aldrich®
Diethylene glycol monomethyl ether
Formula: C₅H₁₂O₃
Boiling Pt: 193 °C (1013 hPa)
Melting Pt: –65 °C
Density: 1.025 g/cm³ (20 °C)
Flash Pt: 91 °C
Storage Temperature: Ambient
MDL Number: MFCD00002871
CAS Number: 111-77-3
EINECS: 203-906-6
Merck Index: 12,06116

Order Now

Specification Test Results

Solubility (20 °C) soluble
Melting Point -65 °C
Molar Mass 120.15 g/mol
Boiling Point 190 - 196 °C (1013 hPa)
Vapor Pressure 0.3 hPa (20 °C)
Flash Point 91 °C
Refractive Index 1.4264 (27 °C, 589 nm)
Explosion Limit 1.6 - 16.1 %(V)
Density 1.018 - 1.022 g/cm3 (20 °C) DIN 51757
pH 4 - 7 (200 g/l, H2O, 20 °C)
Ignition Point 215 °C
Evaporation number 900


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR