Diethylene glycol monoethyl ether for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma

Synonyms: 2-(2-Ethoxyethoxy)ethanol, Di(ethylene glycol) ethyl ether, Diethylene Glycol Monoethyl Ether

M-Clarity™ quality level = MQ200

8.03127.1000 8.03127.2500 8.03127.9025
EM8.03127.1000CS 455.04 CAD
EM8.03127.1000 EM8.03127.2500 CA8.03127.1000 CA8.03127.2500 CA8.03127.9025
Diethylene glycol monoethyl ether for synthesis, Sigma-Aldrich®
Diethylene glycol monoethyl ether
Formula: C₆H₁₄O₃
MW: 134.18 g/mol
Boiling Pt: 197
Melting Pt: -80
Density: 0.99 g/cm³ (20 °C)
Flash Pt: 93
Storage Temperature: Ambient
MDL Number: MFCD00002872
CAS Number: 111-90-0
EINECS: 203-919-7
REACH: 01-2119475105-42
Merck Index: 13,01809

Order Now

Specification Test Results

Solubility (20 °C) soluble
Melting Point -78 °C
Molar Mass 134.17 g/mol
Boiling Point 202 °C (1013 hPa)
Vapor Pressure 0.13 hPa (20 °C)
Flash Point 96 °C
Refractive Index 1.4293 (20 °C, 589 nm)
Explosion Limit 1.8 - 12.2 %(V)
Density 0.99 g/cm3 (20 °C)
Ignition Point 204 °C
Evaporation number 1200


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR