Decahydronaphthalene (mixture of cis and trans isomers) for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

Synonyms: Decalin, Bicyclo[4.4.0]decane, Decahydronaphthalene

M-Clarity™ quality level = MQ200

8.03101.1000 8.03101.0100 8.03101.1000 8.03101.0100
CA8.03101.1000EA 117 CAD
CA8.03101.1000 EM8.03101.0100 EM8.03101.1000 CA8.03101.0100
Decahydronaphthalene (mixture of cis and trans isomers) for synthesis, Sigma-Aldrich®
Decahydronaphthalene (mixture of cis and trans isomers)
Formula: C₁₀H₁₈
MW: 138.25 g/mol
Boiling Pt: 189…191 °C (1013 hPa)
Melting Pt: -40
Density: 0.8696 g/cm³ (20 °C)
Flash Pt: 57
Storage Temperature: Ambient
MDL Number: MFCD00004130
CAS Number: 91-17-8
EINECS: 202-046-9
UN: 1147
ADR: 3 III
Merck Index: 13,02866
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=41b5fff8aa81be0e42af177a8f0e3ac149943c6e&vid=ca9f3c5d71ac621a6216f8b8b28d7cb27b29fd84&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Solubility 0.89 mg/l (25 °C) (experimental)
Melting Point -40 °C
Molar Mass 138.25 g/mol
Boiling Point 189 - 191 °C (1013 hPa)
Vapor Pressure 1.3 hPa (20 °C)
Flash Point 57 °C
Refractive Index 1.4742 (20 °C, 589 nm)
Explosion Limit 0.7 - 4.9 %(V)
Density 0.88 g/cm3 (20 °C)
Ignition Point 255 °C
Viscosity kinematic 2.2 mm2/s (25 °C)
Hazard Class 3
Hazard Class Description Flammable Liquid


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR