N,N-Dimethylformamide dimethyl acetal for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

Synonyms: 1,1-Dimethoxytrimethylamine, Esterate M, N,N-Dimethylformamide Dimethyl Acetal, 1,1-Dimethoxy-N,N-dimethylmethylamine, DMF-DMA

M-Clarity™ quality level = MQ200

8.03068.0100 8.03068.0100 8.03068.0500
EM8.03068.0100CS 3758.82 CAD
EM8.03068.0100 EM8.03068.0500 CA8.03068.0100 CA8.03068.0500
N,N-Dimethylformamide dimethyl acetal for synthesis, Sigma-Aldrich®
N,N-Dimethylformamide dimethyl acetal
Formula: C₅H₁₃NO₂
MW: 119.16 g/mol
Boiling Pt: 102
Density: 0.893 g/cm³ (20 °C)
Flash Pt: 6
MDL Number: MFCD00008482
CAS Number: 4637-24-5
EINECS: 225-063-3

Order Now

Specification Test Results

Solubility (20 °C) (slow decomposition)
Melting Point -85 °C
Molar Mass 119.16 g/mol
Boiling Point 107 - 108 °C
Flash Point 6 °C
Refractive Index 1.3972 (20 °C, 589 nm)
Explosion Limit 1.3 - 17.7 %(V)
Density 0.90 g/cm3 (20 °C)
pH 7 (H2O)
Ignition Point 155 °C
Hazard Class 3
Hazard Class Description Flammable Liquid


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR