2,5-Dimethoxytetrahydrofuran cis- and trans-mixture for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning


M-Clarity™ quality level = MQ200

8.02961.0025 8.02961.0050 8.02961.0250 8.02961.0025 8.02961.0050 8.02961.0250
EM8.02961.0025CS 4681.54 CAD
EM8.02961.0025 EM8.02961.0050 EM8.02961.0250 CA8.02961.0025 CA8.02961.0050 CA8.02961.0250
2,5-Dimethoxytetrahydrofuran cis- and trans-mixture for synthesis, Sigma-Aldrich®
2,5-Dimethoxytetrahydrofuran
Formula: C₆H₁₂O₃
Boiling Pt: 145 °C (1013 hPa)
Melting Pt: –45 °C
Density: 1.021 g/cm³ (20 °C)
Flash Pt: 35 °C
Storage Temperature: Ambient
MDL Number: MFCD00005359
CAS Number: 696-59-3
EINECS: 211-797-1
UN: 1993
ADR: 3,III

Order Now

Specification Test Results

Solubility 300 g/l (20 °C)
Melting Point -45 °C
Molar Mass 132.15 g/mol
Boiling Point 145 - 148 °C (1.013 hPa)
Vapor Pressure 5 hPa (20 °C)
Flash Point 35 °C
Refractive Index 1.418 (20 °C, 589 nm)
Explosion Limit 2.0 - 12.3 %(V)
Density 1.02 g/cm3 (20 °C)
pH 5 (300 g/l, H2O, 20 °C)
Ignition Point 205 °C
Hazard Class 3
Hazard Class Description Flammable Liquid


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR