Bis(2-methoxyethyl)ether stabilised for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

Synonyms: 2-Methoxyethyl ether, 1-Methoxy-2-(2-methoxyethoxy)ethane, Diethylene glycol dimethyl ether, Dimethyldigol, Diglyme

M-Clarity™ quality level = MQ200

8.02934.0250 8.02934.1000 8.02934.2500 8.02934.0250 8.02934.1000 8.02934.2500
EM8.02934.0250CS 1172.36 CAD
EM8.02934.0250 EM8.02934.1000 EM8.02934.2500 CA8.02934.0250 CA8.02934.1000 CA8.02934.2500
Bis(2-methoxyethyl)ether stabilised for synthesis, Sigma-Aldrich®
Bis(2-methoxyethyl) ether
Formula: C₆H₁₄O₃
MW: 134.18 g/mol
Boiling Pt: 162
Melting Pt: -64
Density: 0.944 g/cm³ (20 °C)
Flash Pt: 53
Storage Temperature: Ambient
MDL Number: MFCD00008503
CAS Number: 111-96-6
EINECS: 203-924-4
UN: 3271
ADR: 3 III
Merck Index: 13,03187

Order Now

Specification Test Results

Solubility (20 °C) soluble
Melting Point -64 °C
Molar Mass 134.17 g/mol
Boiling Point 155 - 165 °C (1013 hPa)
Vapor Pressure 2 hPa (20 °C)
Flash Point 70 °C
Refractive Index 1.4097 (20 °C, 589 nm)
Explosion Limit 1.4 - 17.4 %(V)
Density 0.944 g/cm3 (20 °C)
pH (H2O, 20 °C) neutral
Ignition Point 190 °C
Evaporation number 90
Hazard Class 3
Hazard Class Description Flammable Liquid


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR