Bis(2-butoxyethyl)ether for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma

Synonyms: Diethylene glycol dibutyl ether, 2,2-Dibutoxyethyl ether

M-Clarity™ quality level = MQ200

8.02933.0250 8.02933.1000 8.02933.0250 8.02933.1000
EM8.02933.0250CS 1407.78 CAD
EM8.02933.0250 EM8.02933.1000 CA8.02933.0250 CA8.02933.1000
Bis(2-butoxyethyl)ether for synthesis, Sigma-Aldrich®
Bis(2-butoxyethyl) ether
Formula: C₁₂H₂₆O₃
MW: 218.34 g/mol
Boiling Pt: 256
Melting Pt: -60
Density: 0.88 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00009459
CAS Number: 112-73-2
EINECS: 204-001-9

Order Now

Specification Test Results

Solubility 3 g/l (20 °C)
Melting Point -60 °C
Molar Mass 218.33 g/mol
Boiling Point 254 °C (1013 hPa)
Vapor Pressure 0.01 hPa (20 °C)
Flash Point 118 °C
Density 0.88 g/cm3 (20 °C)
Ignition Point 190 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR