Bis(2-ethoxyethyl)ether for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma

Synonyms: 2-Ethoxyethyl ether, Diethyleneglycoldiethylether, Diethylene glycol diethyl ether

M-Clarity™ quality level = MQ200

8.02932.0250 8.02932.1000 8.02932.0250 8.02932.1000
EM8.02932.0250CS 1053.86 CAD
EM8.02932.0250 EM8.02932.1000 CA8.02932.0250 CA8.02932.1000
Bis(2-ethoxyethyl)ether for synthesis, Sigma-Aldrich®
Bis(2-ethoxyethyl) ether
Formula: C₈H₁₈O₃
MW: 162.23 g/mol
Boiling Pt: 189
Melting Pt: -44
Density: 0.91 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00009254
CAS Number: 112-36-7
EINECS: 203-963-7
Merck Index: 13,03145

Order Now

Specification Test Results

Solubility (20 °C) soluble
Melting Point -44 °C
Molar Mass 162.22 g/mol
Boiling Point 189 °C (1013 hPa)
Vapor Pressure 0.5 hPa (20 °C)
Flash Point 71 °C
Refractive Index 1.4115 (20 °C, 589 nm)
Density 0.91 g/cm3 (20 °C)
Ignition Point 174 °C

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR