Dibutyl ether for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
|
Warning
|
Synonyms:
Di-n-butyl ether
M-Clarity™ quality level = MQ200
8.02892.0100
8.02892.1000
8.02892.2500
8.02892.0100
8.02892.1000
8.02892.2500
EM8.02892.0100CS
973.28
CAD
EM8.02892.0100
EM8.02892.1000
EM8.02892.2500
CA8.02892.0100
CA8.02892.1000
CA8.02892.2500
Dibutyl ether for synthesis, Sigma-Aldrich®
Dibutyl ether
Formula:
C₈H₁₈O MW: 130.23 g/mol Boiling Pt: 141 Melting Pt: -95 Density: 0.767 g/cm³ (20 °C) Flash Pt: 25 Storage Temperature: Refrigerator |
MDL Number:
MFCD00009461 CAS Number: 142-96-1 EINECS: 205-575-3 UN: 1149 ADR: 3 III Merck Index: 13,01568 |
Specification Test Results
Solubility | 10 g/l (20 °C) |
Melting Point | -95 °C |
Molar Mass | 130.23 g/mol |
Boiling Point | 140 - 143 °C (1013 hPa) |
Vapor Pressure | 6.4 hPa (20 °C) |
Flash Point | 37 - 38 °C |
Refractive Index | 1.3985 |
Explosion Limit | 0.9 - 8.5 %(V) |
Density | 0.769 g/cm3 (20 °C) |
pH | 5.2 (0.2 g/l, H2O, 20 °C) |
Ignition Point | 175 °C |
Water absorption | 300 mg/kg (20 °C) |
Hazard Class | 3 |
Hazard Class Description | Flammable Liquid |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...