Cyclohexene stabilized for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger


M-Clarity™ quality level = MQ200

8.02824.0100 8.02824.0500 8.02824.1000 8.02824.2500 8.02824.0100 8.02824.0500 8.02824.1000 8.02824.2500
EM8.02824.0100CS 1153.4 CAD
EM8.02824.0100 EM8.02824.0500 EM8.02824.1000 EM8.02824.2500 EM8.02824.5000 CA8.02824.0100 CA8.02824.0500 CA8.02824.1000 CA8.02824.2500 CA8.02824.5000
Cyclohexene stabilized for synthesis, Sigma-Aldrich®
Cyclohexene
Formula: C₆H₁₀
Boiling Pt: 83 °C (1013 hPa)
Melting Pt: –104 °C
Density: 0.81 g/cm³ (20 °C)
Flash Pt: –20 °C
Storage Temperature: Ambient
MDL Number: MFCD00001539
CAS Number: 110-83-8
EINECS: 203-807-8
UN: 2256
ADR: 3,II
Merck Index: 13,02756

Order Now

Specification Test Results

Solubility 0.21 g/l (20 °C)
Melting Point -104 °C
Molar Mass 82.14 g/mol
Boiling Point 83 °C (1013 hPa)
Vapor Pressure 90 hPa (20 °C)
Flash Point 1.2 - 7.7 %(V)
Refractive Index -20 °C DIN 51755 Part 1
Density 0.81 g/cm3 (20 °C)
pH 7 - 8 (0.2 g/l, H2O, 20 °C)
Ignition Point 310 °C
Viscosity kinematic 0.82 mm2/s (calculated)
Hazard Class 3
Hazard Class Description Flammable Liquid


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR