Quinoline for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger


M-Clarity™ quality level = MQ200

8.02407.9049 8.02407.0005 8.02407.1000 8.02407.0005 8.02407.1000
CA8.02407.9049EA 6025 CAD
CA8.02407.9049 EM8.02407.0005 EM8.02407.1000 CA8.02407.0005 CA8.02407.1000
Quinoline for synthesis, Sigma-Aldrich®
Quinoline
Formula: C₉H₇N
MW: 129.16 g/mol
Boiling Pt: 238
Melting Pt: -15
Density: 1.095 g/cm³ (20 °C)
Flash Pt: 107 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00006736
CAS Number: 91-22-5
EINECS: 202-051-6
UN: 2656
ADR: 6.1 III
Merck Index: 13,08160
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e452fdc6c4f70d31bc2cfb5dade041787f8be427&vid=ec752804bedb2e9f8adfbea9ad287c2452f87a17&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Solubility 6 g/l (20 °C)
Melting Point -15 °C
Molar Mass 129.16 g/mol
Boiling Point 237 - 238 °C (1013 hPa)
Vapor Pressure 0.08 hPa (20 °C)
Flash Point 107 °C
Refractive Index 1.6268 (20 °C, 589 nm)
Explosion Limit 1.2 - 7 %(V)
Density 1.09 g/cm3 (20 °C)
pH 7.3 (5 g/l, H2O, 20 °C)
Ignition Point 480 °C
Hazard Class 6.1
Hazard Class Description Toxic Materials


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR