2-Chlorophenol for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
|
Danger
|
Synonyms:
1-Chloro-2-hydroxybenzene
M-Clarity™ quality level = MQ200
8.02253.0100
8.02253.0500
8.02253.2500
8.02253.0100
8.02253.0500
8.02253.2500
EM8.02253.0100CS
911.66
CAD
EM8.02253.0100
EM8.02253.0500
EM8.02253.2500
CA8.02253.0100
CA8.02253.0500
CA8.02253.2500
2-Chlorophenol for synthesis, Sigma-Aldrich®
2-Chlorophenol
Formula:
C₆H₅ClO Boiling Pt: 175 °C (1013 hPa) Melting Pt: 9 °C Density: 1.25 g/cm³ (20 °C) Flash Pt: 72 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002159 CAS Number: 95-57-8 EINECS: 202-433-2 UN: 2021 ADR: 6.1,III Merck Index: 12,02173 |
Specification Test Results
Solubility | 28.5 g/l (20 °C) |
Melting Point | 7 °C |
Molar Mass | 128.55 g/mol |
Boiling Point | 174 °C (1013 hPa) |
Vapor Pressure | 13.3 hPa (50 °C) |
Flash Point | 63.9 °C |
Refractive Index | 1.5579 (20 °C, 589 nm) |
Density | 1.26 g/cm3 (20 °C) |
Ignition Point | 550 °C |
Hazard Class | 6.1 |
Hazard Class Description | Toxic Materials |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...