4-tert-Butylpyrocatechol for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

Synonyms: 4-(tert-Butyl)benzene-1,2-diol, 4-tert-Butylcatechol

M-Clarity™ quality level = MQ200

8.01987.0005 8.01987.0250 8.01987.0005 8.01987.0250
EM8.01987.0005CS 49943.8 CAD
EM8.01987.0005 EM8.01987.0250 CA8.01987.0005 CA8.01987.0250
4-tert-Butylpyrocatechol for synthesis, Sigma-Aldrich®
4-tert-Butylpyrocatechol
Formula: C₁₀H₁₄O₂
MW: 166.22 g/mol
Boiling Pt: 285 °C (1013 hPa)
Melting Pt: 53 °C
Density: 1.05 g/cm³ (60 °C)
Flash Pt: 129 °C
Storage Temperature: Ambient
MDL Number: MFCD00002201
CAS Number: 98-29-3
EINECS: 202-653-9
UN: 2923
ADR: 8,III

Order Now

Specification Test Results

Solubility 2 g/l (20 °C)
Melting Point 52 - 55 °C
Molar Mass 166.22 g/mol
Bulk Density 650 - 700 kg/m3
Boiling Point 284 - 286 °C (1013 hPa)
Vapor Pressure  - 1 hPa (25 °C)
Flash Point 129 °C
Density 1.05 g/cm3 (60 °C)
Ignition Point 160 °C
Viscosity kinematic 100 mm2/s (57 °C)
Hazard Class 8
Hazard Class Description Corrosive Material

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR