Brij® 35 for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
|
Warning
|
Synonyms:
Polyoxyethylene(23)lauryl ether, Tricosaethylene glycol mono-n-dodecyl ether, Brij® L23
M-Clarity™ quality level = MQ200
8.01962.0250
8.01962.1000
8.01962.0250
8.01962.1000
EM8.01962.0250CS
374.46
CAD
EM8.01962.0250
EM8.01962.1000
CA8.01962.0250
CA8.01962.1000
Brij® 35 for synthesis, Sigma-Aldrich®
Brij® 35
Formula:
C₅₈H₁₁₈O₂₄ MW: 1199.56 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00080891 CAS Number: 9002-92-0 EINECS: 500-002-6 |
Specification Test Results
Solubility | (25 °C) soluble |
Melting Point | 36 - 42 °C |
Molar Mass | 582.8 g/mol |
Bulk Density | 1050 kg/m3 |
Boiling Point | 100 °C (1013 hPa) |
Vapor Pressure |
|
Flash Point |
|
Density | 1.05 g/cm3 (20 °C) |
pH | 5.5 - 7.0 (100 g/l, H2O, 20 °C) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...