Butyl vinyl ether stabilised for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

Synonyms: n-Butyl vinyl ether

Stabilization: 100 ppm Stabilised with potassium hydroxide
M-Clarity™ quality level = MQ200

8.01847.0100 8.01847.0500 8.01847.0100 8.01847.0500
EM8.01847.0100CS 1820.16 CAD
EM8.01847.0100 EM8.01847.0500 CA8.01847.0100 CA8.01847.0500
Butyl vinyl ether stabilised for synthesis, Sigma-Aldrich®
Butyl vinyl ether
Formula: C₆H₁₂O
MW: 100.16 g/mol
Boiling Pt: 94
Melting Pt: -91.9
Density: 0.7804 g/cm³ (20 °C)
MDL Number: MFCD00009454
CAS Number: 111-34-2
EINECS: 203-860-7
UN: 2352
ADR: 3 II

Order Now

Specification Test Results

Solubility 3 g/l (20 °C)
Melting Point -113 °C
Molar Mass 100.16 g/mol
Boiling Point 93 - 94 °C (1013 hPa)
Vapor Pressure 51 hPa (20 °C)
Flash Point -6 °C DIN 51755 Part 1
Refractive Index 1.401 (20 °C, 589 nm)
Explosion Limit 1.2 - 8.6 %(V)
Density 0.78 g/cm3 (20 °C)
Ignition Point 190 °C DIN 51794
Hazard Class 3
Hazard Class Description Flammable Liquid


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR