Bromobenzene ≥99% (by GC) for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning

Synonyms: Phenylbromide, Phenyl bromide

M-Clarity™ quality level = MQ200

8.01786.1000 8.01786.0005 8.01786.0250 8.01786.0005 8.01786.0250 8.01786.9026
CA8.01786.1000EA 157 CAD
CA8.01786.1000 EM8.01786.0005 EM8.01786.0250 CA8.01786.0005 CA8.01786.0250 CA8.01786.9026
Bromobenzene ≥99% (by GC) for synthesis, Sigma-Aldrich®
Bromobenzene
Formula: C₆H₅Br
MW: 157.01 g/mol
Boiling Pt: 155
Melting Pt: -31
Density: 1.49 g/cm3 (20 °C)
Flash Pt: 51
Storage Temperature: Ambient
MDL Number: MFCD00000055
CAS Number: 108-86-1
EINECS: 203-623-8
UN: 2514
ADR: 3 III
Merck Index: 13,01390

Order Now

Specification Test Results

Solubility 4 g/l (20 °C)
Melting Point -31 °C
Molar Mass 157 g/mol
Boiling Point 156 °C (1013 hPa)
Vapor Pressure 4 hPa (20 °C)
Flash Point 51 °C
Refractive Index 1.5602 (20 °C, 589 nm)
Explosion Limit 0.5 - 2.5 %(V)
Density 1.49 g/cm3 (20 °C)
Ignition Point 565 °C
Saturation concentration 26 g/m3 (20 °C) Air
Hazard Class 3
Hazard Class Description Flammable Liquid


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR