1-Chlorobutane for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

Synonyms: n-Butyl Chloride

M-Clarity™ quality level = MQ200

8.01640.0100 8.01640.1000 8.01640.2500 8.01640.0100 8.01640.1000 8.01640.2500
EM8.01640.0100CS 936.94 CAD
EM8.01640.0100 EM8.01640.1000 EM8.01640.2500 CA8.01640.0100 CA8.01640.1000 CA8.01640.2500
1-Chlorobutane for synthesis, Sigma-Aldrich®
1-Chlorobutane
Formula: CH₃(CH₂)₃Cl
Boiling Pt: 78.4 °C (1013 hPa)
Melting Pt: –123 °C
Density: 0.8862 g/cm³ (20 °C)
Flash Pt: –17 °C
Storage Temperature: Ambient
MDL Number: MFCD00001009
CAS Number: 109-69-3
EINECS: 203-696-6
UN: 1127
ADR: 3,II
Merck Index: 13,01559
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e2bd980eb6e7ecd91951d46d0ea510119e976dc0&vid=5231549172a609ad96623a80204b11c7c0c69273&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Solubility 0.5 g/l (20 °C)
Melting Point -123 °C
Molar Mass 92.57 g/mol
Boiling Point 79 °C (1013 hPa)
Vapor Pressure 110 hPa (20 °C)
Flash Point -17 °C
Explosion Limit 1.8 - 10.1 %(V)
Density 0.886 g/cm3 (20 °C)
pH (H2O) not applicable
Ignition Point 280 °C
Hazard Class 3
Hazard Class Description Flammable Liquid


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR