Ethyl bromoacetate for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger


M-Clarity™ quality level = MQ200

8.01636.0250 8.01636.1000 8.01636.0250 8.01636.1000
EM8.01636.0250CS 2344.72 CAD
EM8.01636.0250 EM8.01636.1000 CA8.01636.0250 CA8.01636.1000
Ethyl bromoacetate for synthesis, Sigma-Aldrich®
Ethyl bromoacetate
Formula: BrCH₂COOCH₂CH₃
Boiling Pt: 159 °C (1013 hPa)
Melting Pt: –38 °C
Density: 1.5 g/cm³ (20 °C)
Flash Pt: 58 °C
MDL Number: MFCD00000191
CAS Number: 105-36-2
EINECS: 203-290-9
UN: 1603
ADR: 6.1 (3),II

Order Now

Specification Test Results

Solubility (20 °C) decomposes
Melting Point -38 °C
Molar Mass 167.00 g/mol
Boiling Point 158 - 161 °C (1013 hPa)
Vapor Pressure 4.5 hPa (20 °C)
Flash Point 50 °C
Refractive Index 1.451 (20 °C, 589 nm)
Density 1.50 g/cm3 (20 °C)
Hazard Class 6.1
Hazard Class Description Toxic Materials


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR