2-Butoxyethanol for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning

Synonyms: Ethylene glycol butyl ether, Butyl cellosolve, Ethylene glycol monobutyl ether

M-Clarity™ quality level = MQ200

8.01554.1000 8.01554.2500 8.01554.1000 8.01554.2500 8.01554.6025
EM8.01554.1000CS 437.66 CAD
EM8.01554.1000 EM8.01554.2500 CA8.01554.1000 CA8.01554.2500 CA8.01554.6025
2-Butoxyethanol for synthesis, Sigma-Aldrich®
2-Butoxyethanol
Formula: CH₃(CH₂)₃OCH₂CH₂OH
MW: 118.18 g/mol
Boiling Pt: 171
Melting Pt: -70
Density: 0.902 g/cm³ (20 °C)
Flash Pt: 61
Storage Temperature: Ambient
MDL Number: MFCD00002884
CAS Number: 111-76-2
EINECS: 203-905-0
UN: 2810
Merck Index: 13,01558
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ba5a9d2c527d0c167d2248d1936e2d1bd4aeab7a&vid=75260b2608ddc5eb29642178ab193fbcaf54198d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Solubility (20 °C) soluble
Molar Mass 118.17 g/mol
Boiling Point 168 - 170 °C
Vapor Pressure 0.8 hPa (20 °C)
Flash Point 67 °C
Refractive Index 1.4193 (20 °C, 589 nm)
Explosion Limit 1.1 - 10.6 %(V)
Density 0.90 g/cm3 (20 °C)
pH 7 (H2O, 20 °C) (as aqueous solution)
Ignition Point 230 °C
Saturation concentration 5 g/m3 (20 °C) Air
Viscosity kinematic 3.642 mm2/s (20 °C)
Evaporation number 160


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR