Anthrone for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning

Synonyms: Anthracen-9(10H)-one

M-Clarity™ quality level = MQ200

8.01461.0025 8.01461.0100 8.01461.0025 8.01461.0100
EM8.01461.0025CS 25349.52 CAD
EM8.01461.0025 EM8.01461.0100 CA8.01461.0025 CA8.01461.0100
Anthrone for synthesis, Sigma-Aldrich®
Anthrone
Formula: C₁₄H₁₀O
MDL Number: MFCD00001187
CAS Number: 90-44-8
EINECS: 201-994-0

Order Now

Specification Test Results

Solubility 4.6 mg/l (20 °C)
Melting Point 151 - 156 °C
Molar Mass 194.23 g/mol
Bulk Density 450 kg/m3

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR