2-Aminothiazole for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning


M-Clarity™ quality level = MQ200

8.01272.0100 8.01272.0500 8.01272.0100 8.01272.0500
EM8.01272.0100CS 5084.44 CAD
EM8.01272.0100 EM8.01272.0500 CA8.01272.0100 CA8.01272.0500
2-Aminothiazole for synthesis, Sigma-Aldrich®
2-Aminothiazole
Formula: C₃H₄N₂S
Boiling Pt: 216 °C (1013 hPa)
Melting Pt: 86…89 °C
Density: 1.5 g/cm³ (20 °C)
MDL Number: MFCD00005325
CAS Number: 96-50-4
EINECS: 202-511-6
Merck Index: 13,00479

Order Now

Specification Test Results

Solubility 100 g/l (20 °C)
Melting Point 86 - 89 °C
Molar Mass 100.13 g/mol
Bulk Density 600 kg/m3
Boiling Point 117 °C (20 hPa)
Vapor Pressure  - 1 hPa (20 °C)
pH 9.6 (100 g/l, H2O, 20 °C)
Ignition Point 600 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR