Aluminium acetylacetonate for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger


M-Clarity™ quality level = MQ200

8.01059.0025 8.01059.0250 8.01059.0025 8.01059.0250
EM8.01059.0025CS 7991.64 CAD
EM8.01059.0025 EM8.01059.0250 CA8.01059.0025 CA8.01059.0250
Aluminium acetylacetonate for synthesis, Sigma-Aldrich®
Aluminium acetylacetonate
Formula: C₁₅H₂₁AlO₆
Boiling Pt: 315 °C (1013 hPa)
Melting Pt: 192…195 °C
Density: 1.204 g/cm³ (20 °C)
MDL Number: MFCD00000013
CAS Number: 13963-57-0
EINECS: 237-741-6
UN: 3467
ADR: 6.1,II

Order Now

Specification Test Results

Solubility 3 g/l (20 °C)
Melting Point 190 - 193 °C
Molar Mass 324.31 g/mol
Bulk Density 500 kg/m3
Boiling Point 315 °C (1013 hPa)
pH 8 (3 g/l, H2O, 20 °C)
Ignition Point 500 °C
Hazard Class 6.1
Hazard Class Description Toxic Materials


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR