1,2-Dimethoxyethane for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

Synonyms: Dimethyl glycol, Ethylene glycol dimethyl ether, DME

M-Clarity™ quality level = MQ200

8.00856.1000 8.00856.1000 8.00856.2500 8.00856.6010
EM8.00856.1000CS 2088.76 CAD
EM8.00856.1000 CA8.00856.1000 CA8.00856.2500 CA8.00856.6010
1,2-Dimethoxyethane for synthesis, Sigma-Aldrich®
1,2-Dimethoxyethane
Formula: C₄H₁₀O₂
MW: 90.12 g/mol
Boiling Pt: 85
Melting Pt: -69
Density: 1.113 g/cm³ (20 °C)
Flash Pt: –6 °C
Storage Temperature: Ambient
MDL Number: MFCD00008502
CAS Number: 110-71-4
EINECS: 203-794-9
UN: 2252
ADR: 3 II
Merck Index: 13,03251

Order Now

Specification Test Results

Solubility (20 °C) soluble
Melting Point -69 °C
Molar Mass 90.12 g/mol
Boiling Point 84 - 86 °C (1013 hPa)
Vapor Pressure 67 hPa (20 °C)
Flash Point -6 °C
Explosion Limit 1.6 - 10.4 %(V)
Density 0.87 g/cm3 (20 °C)
pH (H2O) neutral
Ignition Point 200 °C
Viscosity kinematic 0.455 mm2/s (25 °C)
Hazard Class 3
Hazard Class Description Flammable Liquid


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR