Ethyl acrylate stabilized for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

Synonyms: Acrylic acid-ethyl ester

Stabilization: Stabilised with hydroquinone monomethyl ether (4-methoxyphenol, MEHQ)
M-Clarity™ quality level = MQ200

8.00836.0100 8.00836.1000
EM8.00836.0100CS 1756.96 CAD
EM8.00836.0100 EM8.00836.1000 CA8.00836.0100 CA8.00836.1000
Ethyl acrylate stabilized for synthesis, Sigma-Aldrich®
Ethyl acrylate
Formula: C₅H₈O₂
MW: 100.12 g/mol
MDL Number: MFCD00009188
CAS Number: 140-88-5
EINECS: 205-438-8
UN: 1917
ADR: 3,II

Order Now

Specification Test Results

Solubility 15 g/l (20 °C)
Melting Point -71 °C
Molar Mass 100.11 g/mol
Boiling Point 100 °C (1013 hPa)
Vapor Pressure 39 hPa (20 °C)
Flash Point 8 °C
Refractive Index 1.4049 (20 °C, 589 nm)
Explosion Limit 1.8 - 14 %(V)
Density 0.92 g/cm3 (20 °C)
Ignition Point 355 °C DIN 51794
Water absorption 124 g/kg (20 °C)
Hazard Class 3
Hazard Class Description Flammable Liquid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR