Isovaleric acid for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
Synonyms:
Isopropyl acetic acid
, 3-Methylbutanoic acid
, 3-Methylbutyric acid
M-Clarity™ quality level = MQ200
8.00820.0250
8.00820.0005
8.00820.1000
8.00820.1000
8.00820.0250
8.00820.0005
EM8.00820.0005CS
2832.23
CAD
EM8.00820.0005
CA8.00820.1000
CA8.00820.0250
EM8.00820.1000
EM8.00820.0250
CA8.00820.0005
Isovaleric acid for synthesis, Sigma-Aldrich®
Isovaleric acid
Formula:
C₅H₁₀O₂ MW: 102.13 g/mol Boiling Pt: 177 °C (1013 hPa) Melting Pt: –33 °C Density: 0.93 g/cm³ (20 °C) Flash Pt: 74 °C |
MDL Number:
MFCD00002726 CAS Number: 503-74-2 UN: 3265 ADR: 8,II Merck Index: 12,05251 |
Specification Test Results
Assay (GC, area%) | ≥ 99.0 % (a/a) |
Density (d 20\u2007°C/ 4\u2007°C) | 0.925 - 0.926 |
Identity (IR) | passes test |
Water (K. F.) | ≤ 0.20 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...