Terephthalic acid for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma


M-Clarity™ quality level = MQ200

8.00762.0100 8.00762.1000 8.00762.0100 8.00762.1000
EM8.00762.0100CS 7615.6 CAD
EM8.00762.0100 EM8.00762.1000 CA8.00762.0100 CA8.00762.1000
Terephthalic acid for synthesis, Sigma-Aldrich®
Terephthalic acid
Formula: C₈H₆O₄
MW: 166.13 g/mol
Boiling Pt: 392 °C (1013 hPa)
Density: 1.472 g/cm³ (20 °C)
Flash Pt: 260 °C
Storage Temperature: Ambient
MDL Number: MFCD00002558
CAS Number: 100-21-0
EINECS: 202-830-0
Merck Index: 12,09306

Order Now

Specification Test Results

Assay (acidimetric) ≥ 98.0 %
Identity (IR) passes test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR