Tetraethyl orthosilicate (TEOS) for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
|
Warning
|
Synonyms:
Tetraethoxysilane
M-Clarity™ quality level = MQ200
8.00658.9050
8.00658.0250
8.00658.1000
8.00658.0250
8.00658.1000
CA8.00658.9050EA
1804
CAD
CA8.00658.9050
EM8.00658.0250
EM8.00658.1000
CA8.00658.0250
CA8.00658.1000
Tetraethyl orthosilicate (TEOS) for synthesis, Sigma-Aldrich®
Tetraethyl orthosilicate (TEOS)
Formula:
Si(OC₂H₅)₄ MW: 208.33 g/mol Boiling Pt: 168 Melting Pt: –85.5 °C Density: 0.933 g/cm³ (20 °C) Flash Pt: 38 Storage Temperature: Ambient |
MDL Number:
MFCD00009062 CAS Number: 78-10-4 EINECS: 201-083-8 UN: 1292 ADR: 3 III Merck Index: 13,03882 |
Specification Test Results
Solubility | (20 °C) insoluble,(slow decomposition) |
Melting Point | -85.5 °C |
Molar Mass | 208.33 g/mol |
Boiling Point | 168.0 °C (1013 hPa) |
Vapor Pressure | 10 hPa (20 °C) |
Flash Point | 37 °C |
Explosion Limit | 1.3 - 23 %(V) |
Density | 0.93 g/cm3 (20 °C) |
pH | (H2O, 20 °C) not applicable |
Ignition Point | 230 °C |
Hazard Class | 3 |
Hazard Class Description | Flammable Liquid |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...