Salicylaldehyde for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning


M-Clarity™ quality level = MQ200

8.00640.0250 8.00640.1000 8.00640.0005 8.00640.0005
CA8.00640.0250EA 59 CAD
CA8.00640.0250 CA8.00640.1000 EM8.00640.0005 CA8.00640.0005
Salicylaldehyde for synthesis, Sigma-Aldrich®
Salicylaldehyde
Formula: C₇H₆O₂
Boiling Pt: 196 °C (1013 hPa)
Melting Pt: –7 °C
Density: 1.17 g/cm³ (20 °C)
Flash Pt: 79 °C
Storage Temperature: Ambient
MDL Number: MFCD00003317
CAS Number: 90-02-8
EINECS: 201-961-0
UN: 3082
ADR: 9,III
Merck Index: 13,08405

Order Now

Specification Test Results

Solubility 5 g/l (20 °C)
Melting Point 224 - 255 °C
Molar Mass 167.12 g/mol
Bulk Density 700 kg/m3
Vapor Pressure  - 0.01 hPa (20 °C)
Flash Point 188 °C
pH 2.0 (5 g/l, H2O, 20 °C)
Ignition Point 620 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR