Pyridine-2,6-dicarboxylic acid for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning

Synonyms: 2,6-Pyridinedicarboxylic acid, Dipicolinic acid, DPA

M-Clarity™ quality level = MQ200

8.00614.0100 8.00614.1000 8.00614.0100 8.00614.1000
EM8.00614.0100CS 17313.64 CAD
EM8.00614.0100 EM8.00614.1000 CA8.00614.0100 CA8.00614.1000
Pyridine-2,6-dicarboxylic acid for synthesis, Sigma-Aldrich®
Pyridine-2,6-dicarboxylic acid
Formula: C₇H₅NO₄
MW: 167.12 g/mol
Boiling Pt: 357
Melting Pt: 327
Flash Pt: 188
Storage Temperature: Ambient
MDL Number: MFCD00006299
CAS Number: 499-83-2
EINECS: 207-894-3

Order Now

Specification Test Results

Solubility 5 g/l (20 °C)
Melting Point 224 - 255 °C
Molar Mass 167.12 g/mol
Bulk Density 700 kg/m3
Vapor Pressure  - 0.01 hPa (20 °C)
Flash Point 188 °C
pH 2.0 (5 g/l, H2O, 20 °C)
Ignition Point 620 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR