Phthalic anhydride zS, Sigma-Aldrich®
Supplier: MilliporeSigma
|
Danger
|
MQ200
8.00592.0100
8.00592.5000
8.00592.0100
8.00592.5000
EM8.00592.0100CS
1014.36
CAD
EM8.00592.0100
EM8.00592.5000
CA8.00592.0100
CA8.00592.5000
Phthalic anhydride zS, Sigma-Aldrich®
Phthalic anhydride
Formula:
C₈H₄O₃ MW: 148.12 g/mol Boiling Pt: 284 Melting Pt: 131 Density: 1.545 g/cm³ (20 °C) Flash Pt: 151 Storage Temperature: Ambient |
MDL Number:
MFCD00005918 CAS Number: 85-44-9 EINECS: 201-607-5 UN: 2214 ADR: 8 III Merck Index: 13,07457 |
Specification Test Results
Solubility | 6 g/l (20 °C) (slow decomposition) |
Melting Point | 131 °C |
Molar Mass | 148.11 g/mol |
Bulk Density | 500 - 700 kg/m3 |
Boiling Point | 285 °C (1013 hPa) |
Vapor Pressure |
|
Flash Point | 152 °C |
Explosion Limit | 1.7 - 10.5 %(V) |
Density | 1.527 g/cm3 (20 °C) |
pH | 2 (6 g/l, H2O, 20 °C) |
Ignition Point | 580 °C |
Hazard Class | 8 |
Hazard Class Description | Corrosive Material |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...