Methyl methacrylate stabilised for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

Synonyms: Methacrylic acid methyl ester

M-Clarity™ quality level = MQ200

8.00590.0100 8.00590.1000 8.00590.2500 8.00590.0100 8.00590.1000 8.00590.2500
EM8.00590.0100CS 876.9 CAD
EM8.00590.0100 EM8.00590.1000 EM8.00590.2500 CA8.00590.0100 CA8.00590.1000 CA8.00590.2500
Methyl methacrylate stabilised for synthesis, Sigma-Aldrich®
Methyl methacrylate (monomer)
Formula: CH₂=C(CH₃)COOCH₃
MW: 100.12 g/mol
Boiling Pt: 100
Melting Pt: -48
Density: 0.93 g/cm³ (20 °C)
Flash Pt: 8
Storage Temperature: Ambient
MDL Number: MFCD00008587
CAS Number: 80-62-6
EINECS: 201-297-1
UN: 1247
ADR: 3 II
REACH: 01-2119452498-28
Merck Index: 12,06005

Order Now

Specification Test Results

Solubility 15 g/l (20 °C)
Melting Point -48 °C
Molar Mass 100.12 g/mol
Boiling Point 101 °C
Vapor Pressure 53 hPa (20 °C)
Flash Point 10 °C
Refractive Index 1.414 (20 °C, 589 nm)
Explosion Limit 2.1 - 12.5 %(V)
Density 0.94 g/cm3 (20 °C)
pH (H2O) not applicable
Ignition Point 430 °C
Hazard Class 3
Hazard Class Description Flammable Liquid


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR