Palmitic acid for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
M-Clarity™ quality level = MQ200
8.00508.0100
8.00508.1000
8.00508.0100
8.00508.1000
EM8.00508.0100CS
1440.96
CAD
EM8.00508.0100
EM8.00508.1000
CA8.00508.0100
CA8.00508.1000
Palmitic acid for synthesis, Sigma-Aldrich®
Palmitic acid
Formula:
C₁₆H₃₂O₂ Boiling Pt: 352 °C (1013 hPa) Melting Pt: 61…63 °C Density: 0.853 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00002747 CAS Number: 57-10-3 EINECS: 200-312-9 Merck Index: 13,07064 |
Specification Test Results
Assay (GC, area%) | ≥ 98 % |
Identity (IR) | passes test |
Melting range( lower value) | ≥ 62 °C |
Melting range( Upper value) | ≤ 64 °C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...