Chloroacetic acid ≥99% (by acidimetry) for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger


M-Clarity™ quality level = MQ200

8.00412.0100 8.00412.1000 8.00412.2500 8.00412.0100 8.00412.1000 8.00412.2500
EM8.00412.0100CS 5397.28 CAD
EM8.00412.0100 EM8.00412.1000 EM8.00412.2500 CA8.00412.0100 CA8.00412.1000 CA8.00412.2500
Chloroacetic acid ≥99% (by acidimetry) for synthesis, Sigma-Aldrich®
Chloroacetic acid
Formula: ClCH₂COOH
Boiling Pt: 188 °C (1013 hPa)
Melting Pt: 63 °C
Density: 1.64 g/cm3 (20 °C)
Flash Pt: 126 °C
Storage Temperature: Ambient
MDL Number: MFCD00002683
CAS Number: 79-11-8
EINECS: 201-178-4
UN: 1751
ADR: 6.1,8,II
Merck Index: 13,02129
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2516e0c26c6c26f4f6f3656bc5b4256e7cf08207&vid=9c9b7ad094469d9089eb2f0ce88275d1423c2b46&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (acidimetric) ≥ 99,0 %(m)
Identity (IR) conforms
Melting range (lower value) ≥ 60 °C
Melting range (upper value) ≤ 65 °C

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR