L(-)-Malic acid for synthesis, for resolution of racemates, Sigma-Aldrich®
Supplier: MilliporeSigma
|
Warning
|
Synonyms:
L(-)-Malic acid, (S)-(-)-Hydroxy-1,4-butanedioic acid, (S)-(-)-Hydroxysuccinic acid, (S)-2-hydroxysuccinic acid, L(-)-Hydroxysuccinic acid, L(-)-Hydroxy-1,4-butanedioic acid, L-(–)-Malic Acid
M-Clarity™ quality level = MQ200
8.00384.0010
8.00384.0050
8.00384.0500
8.00384.1000
8.00384.0010
8.00384.0050
8.00384.0500
8.00384.1000
EM8.00384.0010CS
38280.24
CAD
EM8.00384.0010
EM8.00384.0050
EM8.00384.0500
EM8.00384.1000
CA8.00384.0010
CA8.00384.0050
CA8.00384.0500
CA8.00384.1000
L(-)-Malic acid for synthesis, for resolution of racemates, Sigma-Aldrich®
L(-)-Malic acid
Formula:
C₄H₆O₅ MW: 134.09 g/mol Boiling Pt: 322 Melting Pt: 101…104 °C Density: 1.595 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00064213 CAS Number: 97-67-6 EINECS: 202-601-5 Merck Index: 13,05730 |
Specification Test Results
Assay (acidimetric) | ≥ 99.0 % |
Identity (IR) | passes test |
Melting range (lower value) | ≥ 98 °C |
Melting range (upper value) | ≤ 103 °C |
Spec. rotation α²0/D (c=5 in pyridine) | -30.0 - -27.0 ° |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...