Triethyl citrate for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma

Synonyms: Triethyl 2-hydroxypropane-1,2,3-tricarboxylate, Hydroxytricarballylic acid triethyl ester, Citric acid triethyl ester

M-Clarity™ quality level = MQ200

8.00251.0100 8.00251.0500 8.00251.0100 8.00251.0500
EM8.00251.0100CS 864.26 CAD
EM8.00251.0100 EM8.00251.0500 CA8.00251.0100 CA8.00251.0500
Triethyl citrate for synthesis, Sigma-Aldrich®
Triethyl citrate
Formula: C₁₂H₂₀O₇
Boiling Pt: 294 °C (1013 hPa)
Melting Pt: –55 °C
Density: 1.135 g/cm³ (25 °C)
Flash Pt: 150 °C
MDL Number: MFCD00009201
CAS Number: 77-93-0
EINECS: 201-070-7

Order Now

Specification Test Results

Solubility 65 g/l (25 °C)
Melting Point -55 °C
Molar Mass 276.28 g/mol
Boiling Point 294 °C (1013 hPa)
Vapor Pressure 0.0025 hPa (25 °C)
Flash Point 150 °C
Density 1.135 g/cm3 (25 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR