2-Ethoxyethyl acetate for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

Synonyms: Ethylglycol acetate

M-Clarity™ quality level = MQ200

8.00187.0100 8.00187.0100 8.00187.1000
EM8.00187.0100CS 1123.44 CAD
EM8.00187.0100 CA8.00187.0100 CA8.00187.1000
2-Ethoxyethyl acetate for synthesis, Sigma-Aldrich®
2-Ethoxyethyl acetate
Formula: C₆H₁₂O₃
MW: 132.16 g/mol
Boiling Pt: 156 °C (1013 hPa)
Melting Pt: –62 °C
Density: 0.976 g/cm³ (20 °C)
Flash Pt: 54 °C
MDL Number: MFCD00009251
CAS Number: 111-15-9
UN: 1172
ADR: 3,III
Merck Index: 13,03787

Order Now

Specification Test Results

Solubility 247 g/l (20 °C)
Melting Point -62 °C
Molar Mass 132.15 g/mol
Boiling Point 156 °C (1013 hPa)
Vapor Pressure 2.67 hPa (20 °C)
Flash Point 52 °C
Refractive Index 1.4068 (20 °C, 589 nm)
Explosion Limit 1.3 - 7.2 %(V)
Density 0.97 g/cm3 (20 °C)
pH 4 - 5 (H2O, 20 °C) (saturated solution)
Ignition Point 379 °C
Saturation concentration 14 g/m3 (20 °C) Air
Evaporation number 47
Hazard Class 3
Hazard Class Description Flammable Liquid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR