Benzilic acid for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma


M-Clarity™ quality level = MQ200

8.00140.0250 8.00140.1000 8.00140.0250 8.00140.1000
EM8.00140.0250CS 1714.3 CAD
EM8.00140.0250 EM8.00140.1000 CA8.00140.0250 CA8.00140.1000
Benzilic acid for synthesis, Sigma-Aldrich®
Benzilic acid
Formula: C₁₄H₁₂O₃
MW: 228.25 g/mol
Boiling Pt: 409
Melting Pt: 150…152 °C
Storage Temperature: Ambient
MDL Number: MFCD00004447
CAS Number: 76-93-7
EINECS: 200-993-2
Merck Index: 13,01080

Order Now

Specification Test Results

Assay (acidimetric)  ≥ 98.0 %
Melting range (lower value)  ≥ 146 °C
Melting range (upper value)  ≤ 150 °C
Identity (IR) passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR