Ammonium carbonate, EMSURE® ACS, Reag. Ph. Eur. for analysis, Supelco®
Supplier: MilliporeSigma
|
Danger
|
M-Clarity™ quality level = MQ300
1.59504.0250
1.59504.1000
CA1.59504.0250EA
131
CAD
CA1.59504.0250
CA1.59504.1000
Ammonium carbonate, EMSURE® ACS, Reag. Ph. Eur. for analysis, Supelco®
Ammonium carbonate
Formula:
(NH₄)₂CO₃ Melting Pt: 58 °C Density: 1.5 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00010890 CAS Number: 506-87-6 EINECS: 208-058-0 Merck Index: 13,00508 |
Specification Test Results
Assay (acidimetric, NH₃) | ≥ 30.0 % |
Identity | passes test |
Description | white crystals |
Insoluble matter | ≤ 0.005 % |
Chloride (Cl) | ≤ 0.0005 % |
Sulfur compounds (as SO₄) | ≤ 0.002 % |
Heavy metals (as Pb) | ≤ 0.0005 % |
Fe (Iron) | ≤ 0.0005 % |
Non-volatile matter | ≤ 0.01 % |
Conforms ACS, Reag. Ph. Eur. | passes test |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...