Methylene blue (C.I. 52015), Supelco®

Supplier: MilliporeSigma
Warning

Synonyms: Basic Blue 9, Methylthioninium chloride, Methylene blue chloride, 3,7-Bis(dimethylamino)phenothiazin-5-ium chloride, Swiss Blue

M-Clarity™ quality level = MQ200

1.59270.0010 1.59270.0100 1.59270.0010 1.59270.0100
EM1.59270.0010CS 4245.46 CAD
EM1.59270.0010 EM1.59270.0100 CA1.59270.0010 CA1.59270.0100
Methylene blue (C.I. 52015), Supelco®
Methylene blue
Formula: C₁₆H₁₈ClN₃S
MW: 319.86 g/mol
Melting Pt: 180
Density: 1.757 g/cm³ (22 °C)
Storage Temperature: Ambient
MDL Number: MFCD00150008
CAS Number: 61-73-4
EINECS: 200-515-2
Merck Index: 12,06137

Order Now

Specification Test Results

Solubility 50 g/l (20 °C)
Melting Point 180 °C (decomposition)
Molar Mass 319.86 g/mol (anhydrous)
Bulk Density 400 - 600 kg/m3
pH 3 (10 g/l, H2O, 20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR