Extran® AP17 liquid, alkaline concentrate

Supplier: MilliporeSigma
1.40006.2500 1.40006.9010 1.40006.9025 1.40006.5000
CA1.40006.2500CS 460 CAD
CA1.40006.2500 CA1.40006.9010 CA1.40006.9025 CA1.40006.5000
Extran® AP17 liquid, alkaline concentrate
Cleaning Agents Detergents and Surface Cleansers
Extran® AP 17 is an active universal cleaning agent for the main wash cycle which cleans and removes even heavily soiled items. Particularly suitable for the removal of starch and protein residues. Extran® AP 17 alkaline does not foam even during heavy agitation in a washing machine.

  • Liquid
  • Free from surfactants
  • Alkaline
  • Chlorine-free and phosphate-free
  • Free from odorants/dyestuffs

Extran® AP 17 is free from organic sufactants and emulsifiers, but contains complexing agents and can therefore be used in both soft and hard water.

Dosing
The idealMore Product Information
Order Now

SPECIFICATIONS

Environmentally Preferable
Formulation Complexing agent, sodium hydroxide solution
Specific gravity 1.294 g/cm³ at 20 °C
Storage temperature 15…25 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR